Merck
  • Antagonistic and synergistic effects of light irradiation on the effects of copper on Chlamydomonas reinhardtii.
Antagonistic and synergistic effects of light irradiation on the effects of copper on Chlamydomonas reinhardtii.
Aquatic toxicology (Amsterdam, Netherlands) (2014-07-30)
Giulia Cheloni, Claudia Cosio, Vera I Slaveykova
ABSTRACT
The present study showed the important role of light intensity and spectral composition on Cu uptake and effects on green alga Chlamydomonas reinhardtii. High-intenisty light (HL) increased cellular Cu concentrations, but mitigated the Cu-induced decrease in chlorophyll fluorescence, oxidative stress and lipid peroxidation at high Cu concentrations, indicating that Cu and HL interact in an antagonistic manner. HL up-regulated the transcription of genes involved in the antioxidant response in C. reinhardtii and thus reduced the oxidative stress upon exposure to Cu and HL. Combined exposure to Cu and UVBR resulted in an increase of cellular Cu contents and caused severe oxidative damage to the cells. The observed effects were higher than the sum of the effects corresponding to exposure to UVBR or Cu alone suggesting a synergistic interaction.
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Copper, tube, 1000mm, outside diameter 1.2mm, inside diameter 0.56mm, wall thickness 0.32mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 12.7mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 5m coil, thickness 0.05mm, coil width 300mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.075mm, half hard, 99.95+%
Copper, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.075mm, half hard, 99.95+%
Copper, tube, 500mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.125mm, hard, 99.95+%
Copper, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.99+%
Copper, tube, 500mm, outside diameter 2.5mm, inside diameter 1.9mm, wall thickness 0.3mm, as drawn, 99.9%
Copper, tube, 200mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 2m coil, thickness 0.20mm, hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.25mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.075mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.05mm, hard, 99.95+%
Copper, wire reel, 100m, diameter 0.5mm, as drawn, 99.98+%
Copper - O.F.H.C., foil, 6mm disks, thickness 0.125mm, hard, 99.95+%