Merck
  • Root biomass production in populations of six rooted macrophytes in response to Cu exposure: intra-specific variability versus constitutive-like tolerance.
Root biomass production in populations of six rooted macrophytes in response to Cu exposure: intra-specific variability versus constitutive-like tolerance.
Environmental pollution (Barking, Essex : 1987) (2014-07-25)
L Marchand, F Nsanganwimana, J B Lamy, C Quintela-Sabaris, C Gonnelli, I Colzi, T Fletcher, N Oustrière, A Kolbas, P Kidd, F Bordas, P Newell, P Alvarenga, A Deletic, M Mench
ABSTRACT
Intra-specific variability of root biomass production (RP) of six rooted macrophytes, i.e. Juncus effusus, Phragmites australis, Schoenoplectus lacustris, Typha latifolia, Phalaris arundinacea, and Iris pseudacorus grown from clones, in response to Cu exposure was investigated. Root biomass production varied widely for all these macrophytes in control conditions (0.08 μM) according to the sampling site. Root biomass production of T. latifolia and I. pseudacorus in the 2.5-25 μM Cu range depended on the sampling location but not on the Cu dose in the growth medium. For P. australis, J. effusus, S. lacustris, and P. arundinacea, an intra-specific variability of RP depending on both the sampling location and the Cu-dose was evidenced. This intra-specific variability of RP depending on the sampling location and of Cu-tolerance for these last four species suggests that Cu constitutive tolerance for all rooted macrophytes is not a species-wide trait but it exhibits variability for some species.
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Copper - O.F.H.C., foil, 5m coil, thickness 0.075mm, half hard, 99.95+%
Copper, wire reel, 500m, diameter 0.125mm, annealed, 99.9%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.05mm, hard, 99.95+%
Copper - O.F.H.C., foil, 2m coil, thickness 0.05mm, hard, 99.95+%
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Copper - O.F.H.C., foil, 6mm disks, thickness 0.006mm, hard, 99.95+%
Copper - O.F.H.C., foil, 50m coil, thickness 0.05mm, coil width 10mm, annealed, 99.95+%
Copper - O.F.H.C., foil, 5m coil, thickness 0.125mm, hard, 99.95+%
Copper, wire reel, 100m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Copper, wire reel, 100m, diameter 0.1mm, annealed, 99.9%
Copper, wire reel, 1m, diameter 0.01mm, hard, 99.99%
Copper, wire reel, 500m, diameter 0.25mm, annealed, 99.9%
Copper, wire reel, 50m, diameter 0.15mm, annealed, silver plated, 99.9%
Copper, wire reel, 10m, diameter 0.02mm, annealed, 99.99%
Copper, wire reel, 200m, diameter 0.5mm, annealed, silver plated, 99.9%
Copper, wire reel, 5000m, diameter 0.04mm, annealed, 99.9%
Copper, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Copper, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.99+%
Copper, wire reel, 500m, diameter 0.1mm, annealed, 99.9%