Merck
  • METT-10, a putative methyltransferase, inhibits germ cell proliferative fate in Caenorhabditis elegans.
METT-10, a putative methyltransferase, inhibits germ cell proliferative fate in Caenorhabditis elegans.
Genetics (2009-07-15)
Maia Dorsett, Bethany Westlund, Tim Schedl
ABSTRACT
Germ-line stem cells are unique because they either self-renew through mitosis or, at a certain frequency, switch to meiosis and produce gametes. The switch from proliferation to meiosis is tightly regulated, and aberrations in switching result in either too little or too much proliferation. To understand the genetic basis of this regulation, we characterized loss-of-function mutations and a novel tumorous allele of Caenorhabditis elegans mett-10, which encodes a conserved putative methyltransferase. We show that METT-10 is a nuclear protein that acts in the germ line to inhibit the specification of germ-cell proliferative fate. METT-10 also promotes vulva, somatic gonad, and embryo development and ensures meiotic development of those germ cells that do differentiate. In addition, phenotypic analysis of a mett-10 null allele reveals that METT-10 enables mitotic cell cycle progression. The finding that METT-10 functions to inhibit germ-cell proliferative fate, despite promoting mitotic cell cycle progression of those germ cells that do proliferate, separates the specification of proliferative fate from its execution.
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for FC IV, sufficient for 50 assays, Kit size L
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for FC IV, sufficient for 50 assays, Kit size M
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC IV, sufficient for 50 assays, Kit size M
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC IV, sufficient for 50 assays, Kit size S
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for HTS IV, sufficient for 200 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for HTS IV, sufficient for 200 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 594 for FC IV, sufficient for 50 assays, Kit size S
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for FC IV, sufficient for 50 assays, Kit size S
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for FC IV size M, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for FC IV, sufficient for 50 assays, Kit size L
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for FC IV size M, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 555 for FC IV size M, sufficient for 50 assays
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for HTS IV, sufficient for 200 assays, Kit size M
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for HTS IV, sufficient for 200 assays, Kit size S
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 647 for FC IV, sufficient for 50 assays, Kit size M