Merck
Silicon: the health benefits of a metalloid.
Metal ions in life sciences (2014-01-29)
Keith R Martin
ABSTRACT
Silicon is the second most abundant element in nature behind oxygen. As a metalloid, silicon has been used in many industrial applications including use as an additive in the food and beverage industry. As a result, humans come into contact with silicon through both environmental exposures but also as a dietary component. Moreover, many forms of silicon, that is, Si bound to oxygen, are water-soluble, absorbable, and potentially bioavailable to humans presumably with biological activity. However, the specific biochemical or physiological functions of silicon, if any, are largely unknown although generally thought to exist. As a result, there is growing interest in the potential therapeutic effects of water-soluble silica on human health. For example, silicon has been suggested to exhibit roles in the structural integrity of nails, hair, and skin, overall collagen synthesis, bone mineralization, and bone health and reduced metal accumulation in Alzheimer's disease, immune system health, and reduction of the risk for atherosclerosis. Although emerging research is promising, much additional, corroborative research is needed particularly regarding speciation of health-promoting forms of silicon and its relative bioavailability. Orthosilicic acid is the major form of bioavailable silicon whereas thin fibrous crystalline asbestos is a health hazard promoting asbestosis and significant impairment of lung function and increased cancer risk. It has been proposed that relatively insoluble forms of silica can also release small but meaningful quantities of silicon into biological compartments. For example, colloidal silicic acid, silica gel, and zeolites, although relatively insoluble in water, can increase concentrations of water-soluble silica and are thought to rely on specific structural physicochemical characteristics. Collectively, the food supply contributes enough silicon in the forms aforementioned that could be absorbed and significantly improve overall human health despite the negative perception of silica as a health hazard. This review discusses the possible biological potential of the metalloid silicon as bioavailable orthosilicic acid and the potential beneficial effects on human health.
Product Description
Silicon, rod, 100mm, diameter 12.7mm, single crystal - random orientation, 100%
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains phosphorus as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer, <111>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.3 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, P-type, contains boron as dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains phosphorus as dopant, diam. × thickness 3 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <111>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, wafer (single side polished), <100>, N-type, contains no dopant, diam. × thickness 2 in. × 0.5 mm
Sigma-Aldrich
Silicon, nanopowder, <100 nm particle size (TEM), ≥98% trace metals basis
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Sigma-Aldrich
Silicon, powder, −325 mesh, 99% trace metals basis
Sigma-Aldrich
Silicon, pieces, 99.95% trace metals basis
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%