Merck
  • Evaluation of 5-fluorouracil pharmacokinetic models and therapeutic drug monitoring in cancer patients.
Evaluation of 5-fluorouracil pharmacokinetic models and therapeutic drug monitoring in cancer patients.
Pharmacogenomics (2013-05-09)
André Bp van Kuilenburg, Jan Gerard Maring
ABSTRACT
5-fluorouracil (5-FU) remains the cornerstone of all currently applied regimens for the treatment of patients with cancers of the gastrointestinal tract, breast, and head and neck. Unfortunately, a large variation in the clearance of 5-FU has been observed between patients, suggesting that some patients might receive nonoptimal 5-FU doses. However, therapeutic drug monitoring of 5-FU has been shown to result in reduced intra- and inter-individual variability in 5-FU plasma levels and pharmacokinetically guided dose adjustments of 5-FU-containing therapy results in a significantly improved efficacy and tolerability. To date, compartmental Michaelis-Menten elimination-based modeling has proven to be a sensitive and accurate tool for analyzing the pharmacokinetics of 5-FU and to identify patients with a dihydropyrimidine dehydrogenase deficiency. These Michaelis-Menten models also allow the use of a limited sampling strategy and offer the opportunity to predict a priori the 5-FU plasma concentrations in patients receiving adapted doses of 5-FU.
Product Description
Supelco
5-Fluorouracil, analytical standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Fluorouracil, European Pharmacopoeia (EP) Reference Standard