Merck
Zirconium for esthetic rehabilitation: an overview.
Indian journal of dental research : official publication of Indian Society for Dental Research (2011-04-29)
Anjana Raut, P Laxman Rao, T Ravindranath
ABSTRACT
The demand for esthetic restorations has resulted in an increased use of dental ceramics for anterior and posterior restorations. A few decades ago, all-ceramic restorations were restricted to treatment in the anterior region, but now all-ceramic restorations can be made anywhere in the dentition. The properties of traditional ceramic materials, however, have limited their use to single crowns; larger restorations have been inadvisable because of insufficient strength. In attempts to meet the requirements for dental materials and improve strength and toughness, several new ceramic materials and techniques have been developed during the past few decades The paper reviews the current literature on dental zirconia with respect to survival, properties, marginal fit, cementation, esthetics and suggests clinical recommendations for their use.
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Zirconium, foil, not light tested, 100x100mm, thickness 0.006mm, 99.2%
Zirconium, foil, light tested, 150x150mm, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.016mm, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.2%
Zirconium, foil, not light tested, 25x25mm, thickness 0.001mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.008mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.075mm, annealed, 99.2%
Zirconium, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.05mm, annealed, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.01mm, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.004mm, 99.2%
Zirconium, foil, not light tested, 50x50mm, thickness 0.003mm, 99.2%
Zirconium, foil, light tested, 25x25mm, thickness 0.015mm, 99.2%
Zirconium, foil, light tested, 100x300mm, thickness 0.016mm, 99.2%
Zirconium, foil, light tested, 50x50mm, thickness 0.007mm, 99.2%
Zirconium, foil, light tested, 200x300mm, thickness 0.075mm, annealed, 99.2%
Zirconium, wire reel, 1m, diameter 0.05mm, as drawn, 99.2%
Zirconium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.55mm, wall thickness 0.9mm, annealed, 99.2%
Zirconium, sponge, 200g, max. size 25mm, 99.2%
Zirconium, sponge, 500g, max. size 25mm, 99.2%
Zirconium, tube, 1000mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, annealed, 99.2%
Zirconium, tube, 500mm, outside diameter 2.0mm, inside diameter 1.6mm, wall thickness 0.2mm, annealed, 99.2%
Zirconium, rod, 50mm, diameter 19.0mm, centerless ground, 99.2%
Zirconium, rod, 200mm, diameter 3.0mm, centerless ground, 99.2%
Zirconium, wire reel, 2m, diameter 0.05mm, as drawn, 99.2%