Merck
Carbamazepine for acute and chronic pain in adults.
The Cochrane database of systematic reviews (2011-01-21)
Philip J Wiffen, Sheena Derry, R Andrew Moore, Henry J McQuay
ABSTRACT
Carbamazepine is used to treat chronic neuropathic pain. Evaluation of analgesic efficacy and adverse effects of carbamazepine for acute and chronic pain management (except headaches). Randomised controlled trials (RCTs) of carbamazepine in acute, chronic or cancer pain were identified, searching MEDLINE, EMBASE, SIGLE and Cochrane CENTRAL to June 2010, reference lists of retrieved papers, and reviews. RCTs reporting the analgesic effects of carbamazepine. Two authors independently extracted results and scored for quality. Numbers needed to treat to benefit (NNT) or harm (NNH) with 95% confidence intervals (CI) were calculated from dichotomous data for effectiveness, adverse effects and adverse event withdrawal. Issues of study quality, size, duration, and outcomes were examined. Fifteen included studies (12 cross-over design; three parallel-group) with 629 participants.Carbamazepine was less effective than prednisolone in preventing postherpetic neuralgia following acute herpes zoster (1 study, 40 participants). No studies examined acute postoperative pain.Fourteen studies investigated chronic neuropathic pain: two lasted eight weeks, others were four weeks or less (mean 3 weeks, median 2 weeks). Five had low reporting quality. Ten involved fewer than 50 participants; mean and median maximum treatment group sizes were 34 and 29. Outcome reporting was inconsistent.Most placebo controlled studies indicated that carbamazepine was better than placebo. Five studies with 298 participants provided dichotomous results; 70% improved with carbamazepine and 12% with placebo. Carbamazepine at any dose, using any definition of improvement was significantly better than placebo (70% versus 12% improved; 5 studies, 298 participants); relative benefit 6.1 (3.9 to 9.7), NNT 1.7 (1.5 to 2.0). Four studies (188 participants) reporting outcomes equivalent to 50% pain reduction or more over baseline had a similar NNT.With carbamazepine, 66% of participants experienced at least one adverse event, and 27% with placebo; relative risk 2.4 (1.9 to 3.1), NNH 2.6 (2.1 to 3.5). Adverse event withdrawals occurred in12 of 323 participants (4%) with carbamazepine and 0 of 310 with placebo. Serious adverse events were not reported consistently; rashes were associated with carbamazepine. Five deaths occurred in patients on carbamazepine, with no obvious drug association. Carbamazepine is effective in chronic neuropathic pain, with caveats. No trial was longer than four weeks, of good reporting quality, using outcomes equivalent to at least moderate clinical benefit. In these circumstances, caution is needed in interpretation, and meaningful comparison with other interventions is not possible.
Product Description
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Carbamazepine, powder
Supelco
Carbamazepine, analytical standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard