Merck
  • Amino acid side chain interaction with chelate-liganded crosslinked dextran, agarose and TSK gel. A mini review of recent work.
Amino acid side chain interaction with chelate-liganded crosslinked dextran, agarose and TSK gel. A mini review of recent work.
Journal of molecular recognition : JMR (1990-06-01)
J Porath
ABSTRACT
Protein adsorption and retention data collected from recent chromatographic studies on hydrophilic gels substituted with chelate-bonded metal ions are discussed. Attempts are made to interpret the adsorption behavior in terms of molecular events caused by the affinity for the immobilized metal ions.
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, for molecular biology
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L