Merck
  • Iodine transfers in the coastal marine environment: the key role of brown algae and of their vanadium-dependent haloperoxidases.
Iodine transfers in the coastal marine environment: the key role of brown algae and of their vanadium-dependent haloperoxidases.
Biochimie (2006-09-30)
Catherine Leblanc, Carole Colin, Audrey Cosse, Ludovic Delage, Stéphane La Barre, Pascal Morin, Bruno Fiévet, Claire Voiseux, Yves Ambroise, Elodie Verhaeghe, David Amouroux, Olivier Donard, Emmanuel Tessier, Philippe Potin
ABSTRACT
Brown algal kelp species are the most efficient iodine accumulators among all living systems, with an average content of 1.0% of dry weight in Laminaria digitata, representing a ca. 30,000-fold accumulation of this element from seawater. Like other marine macroalgae, kelps are known to emit volatile short-lived organo-iodines, and molecular iodine which are believed to be a main vector of the iodine biogeochemical cycle as well as having a significant impact on atmospheric chemistry. Therefore, radioactive iodine can potentially accumulate in seaweeds and can participate in the biogeochemical cycling of iodine, thereby impacting human health. From a radioecological viewpoint, iodine-129 (129I, half-life of 1.6 x 10(7) years) is one of the most persistent radionuclide released from nuclear facilities into the environment. In this context, the speciation of iodine by seaweeds is of special importance and there is a need to further understand the mechanisms of iodine uptake and emission by kelps. Recent results on the physiological role and biochemistry of the vanadium haloperoxidases of brown algae emphasize the importance of these enzymes in the control of these processes.
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%