Merck
Macrolides and ketolides: azithromycin, clarithromycin, telithromycin.
Infectious disease clinics of North America (2004-08-17)
Jerry M Zuckerman
ABSTRACT
The advanced macrolides, azithromycin and clarithromycin, and the ketolide telithromycin are structural analogues of erythromycin. They have several distinct advantages when compared with erythromycin including enhanced spectrum of activity, more favorable pharmacokinetics and pharmacodynamics, once daily administration, and improved tolerability. This article reviews the pharmacokinetics, antimicrobial activity, clinical use, and adverse effects of these antimicrobial agents.
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Supelco
Clarithromycin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Clarithromycin, United States Pharmacopeia (USP) Reference Standard
Clarithromycin, European Pharmacopoeia (EP) Reference Standard
Clarithromycin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
USP
Clarithromycin Identity, United States Pharmacopeia (USP) Reference Standard