Merck
  • Impact of cellular metabolism on the biological effects of benzo[a]pyrene and related hydrocarbons.
Impact of cellular metabolism on the biological effects of benzo[a]pyrene and related hydrocarbons.
Drug metabolism reviews (2001-03-29)
K P Miller, K S Ramos
ABSTRACT
Polycyclic aromatic hydrocarbons are ubiquitous contaminants in the environment. Benzo[a]pyrene (BaP), a prototypical member of this class of chemicals, has been extensively studied for its toxic effects in laboratory animals and human populations. BaP toxicity is often mediated by oxidative metabolism to reactive intermediates that interact with macromolecules leading to alterations in target cell structure and function. More recent evidence suggests that disruption of cellular signaling pathways involved in the regulation of growth and differentiation contribute significantly to the toxicity of BaP and its metabolites. This review summarizes recent advances in our understanding of biological mechanisms of BaP toxicity at the molecular level, and the role of metabolic intermediates in carcinogenesis, atherogenesis, and teratogenesis.
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone