Merck
  • Global rates of marine sulfate reduction and implications for sub-sea-floor metabolic activities.
Global rates of marine sulfate reduction and implications for sub-sea-floor metabolic activities.
Science (New York, N.Y.) (2014-05-09)
Marshall W Bowles, José M Mogollón, Sabine Kasten, Matthias Zabel, Kai-Uwe Hinrichs
ABSTRACT
Sulfate reduction is a globally important redox process in marine sediments, yet global rates are poorly quantified. We developed an artificial neural network trained with 199 sulfate profiles, constrained with geomorphological and geochemical maps to estimate global sulfate-reduction rate distributions. Globally, 11.3 teramoles of sulfate are reduced yearly (~15% of previous estimates), accounting for the oxidation of 12 to 29% of the organic carbon flux to the sea floor. Combined with global cell distributions in marine sediments, these results indicate a strong contrast in sub-sea-floor prokaryote habitats: In continental margins, global cell numbers in sulfate-depleted sediment exceed those in the overlying sulfate-bearing sediment by one order of magnitude, whereas in the abyss, most life occurs in oxic and/or sulfate-reducing sediments.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Millipore
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Carbon, foil, 5x5mm, thickness 2.0mm, hOpg
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Carbon, foil, 12mm disks, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, tube, graphite, 50mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, rod, 300mm, diameter 13.0mm, graphite, 100%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 300mm, diameter 10.0mm, graphite, 100%
Carbon, rod, 100mm, diameter 13.0mm, graphite, 100%
Carbon, rod, 50mm, diameter 25mm, graphite, 100%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon - Vitreous, rod, 200mm, diameter 10mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%