Merck
  • Synthesis and characterization of CoFe2O4/polyaniline nanocomposites for electromagnetic interference applications.
Synthesis and characterization of CoFe2O4/polyaniline nanocomposites for electromagnetic interference applications.
Journal of nanoscience and nanotechnology (2014-04-18)
K Praveena, S Srinath
ABSTRACT
The Cobalt ferrite (CoFe2O4) powders were synthesized by Co-precipitation method. The as prepared ferrite powders were incorporated into a polyaniline matrix at various volumetric ratios. The as prepared composites of ferrite and polyaniline powders were characterized using X-ray diffraction (XRD), transmission electron microscope (TEM). The particle size of CoFe2O4 is found to be 20 nm. The saturation magnetization (M(s)) of all the composites was found to be decreasing with decrease of ferrite content, while coercivity (H(c)) remained at the value corresponding to pure cobalt ferrite nanopowders. The complex permittivity (epsilon' and epsilon") and permeability (mu' and mu") of composite samples were measured in the range of 1 MHz to 1.1 GHz. The value of epsilon' and mu' found to be increased with ferrite volume concentration.
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Cobalt, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.006mm, 99.9%
Cobalt, foil, not light tested, 25x25mm, thickness 0.007mm, 99.9%
Cobalt, foil, not light tested, 50x50mm, thickness 0.008mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, wire reel, 0.1m, diameter 0.1mm, hard, 99.99+%
Cobalt, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Cobalt, microfoil, disks, 25mm, thinness 0.5μm, specific density 434μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, microfoil, disks, 25mm, thinness 0.25μm, specific density 222μg/cm2, permanent mylar 3.5μm support, 99.9%
Cobalt, wire reel, 1m, diameter 0.125mm, hard, 99.99+%
Cobalt, wire reel, 2m, diameter 0.25mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 0.25mm, hard, 99.99+%
Cobalt, wire reel, 0.5m, diameter 1.0mm, hard, 99.99+%
Cobalt, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Cobalt, wire reel, 0.1m, diameter 0.05mm, hard, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.125mm, hard, 99.99+%
Cobalt, foil, 250x250mm, thickness 0.50mm, annealed, 99.9%
Cobalt, foil, 15mm disks, thickness 0.025mm, 99.9%