Merck
  • Efficient infrared luminescence of Er2S3/ZnS core/shell quantum dots and IR QDs-lED.
Efficient infrared luminescence of Er2S3/ZnS core/shell quantum dots and IR QDs-lED.
Journal of nanoscience and nanotechnology (2014-04-18)
Bo Li, Xiaosong Zhang, Ting Ji, Gaofeng Zhang, Lan Li
ABSTRACT
We have synthesized Er2S3/ZnS core/shell QDs by employing ErSt3, ZnSt2, and sulfur as precursors via a hot solution phase chemistry using a nucleation-doping strategy. X-ray diffraction (XRD), transmission electron microscope (TEM) and photoluminescence (PL) spectra were used to characterize the structure, morphology and luminescence properties of Er2S3/ZnS core/shell QDs. Moreover, the influence of overcoating temperatures on the infrared luminescence properties of QDs was investigated. PL spectra show that the emission intensity from the 4I13/2 --> 4I15/2 transition of Er3+ strongly increases with increasing overcoating temperatures, which was interpreted by the enhancing diffusion of Er3+ ions. IR-LEDs were fabricated combining commercial red GaAs LEDs with Er2S3/ZnS QDs, and luminescence properties of the IR-LED have been investigated.
Sigma-Aldrich
Zinc selenide, powder, 10 μm, 99.99% trace metals basis
Erbium, rod, 100mm, diameter 10.0mm, cast, 99%
Erbium, foil, 25x25mm, thickness 1.0mm, as rolled, 99%
Erbium, rod, 50mm, diameter 6.35mm, cast, 99%
Erbium, foil, 50x50mm, thickness 0.125mm, as rolled, 99%
Erbium, rod, 50mm, diameter 2.0mm, cast, 99%
Erbium, foil, 50x50mm, thickness 1.0mm, as rolled, 99%
Erbium, rod, 100mm, diameter 6.35mm, cast, 99%
Erbium, wire reel, 50mm, diameter 1.0mm, hard, 99.9%
Erbium, wire reel, 100mm, diameter 1.0mm, hard, 99.9%
Erbium, rod, 50mm, diameter 10.0mm, cast, 99%
Erbium, foil, 25x25mm, thickness 0.125mm, as rolled, 99%
Erbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Erbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%