Merck
  • Distinct phenotype of a Wilson disease mutation reveals a novel trafficking determinant in the copper transporter ATP7B.
Distinct phenotype of a Wilson disease mutation reveals a novel trafficking determinant in the copper transporter ATP7B.
Proceedings of the National Academy of Sciences of the United States of America (2014-04-08)
Lelita T Braiterman, Amrutha Murthy, Samuel Jayakanthan, Lydia Nyasae, Eric Tzeng, Grazyna Gromadzka, Thomas B Woolf, Svetlana Lutsenko, Ann L Hubbard
ABSTRACT
Wilson disease (WD) is a monogenic autosomal-recessive disorder of copper accumulation that leads to liver failure and/or neurological deficits. WD is caused by mutations in ATP7B, a transporter that loads Cu(I) onto newly synthesized cupro-enzymes in the trans-Golgi network (TGN) and exports excess copper out of cells by trafficking from the TGN to the plasma membrane. To date, most WD mutations have been shown to disrupt ATP7B activity and/or stability. Using a multidisciplinary approach, including clinical analysis of patients, cell-based assays, and computational studies, we characterized a patient mutation, ATP7B(S653Y), which is stable, does not disrupt Cu(I) transport, yet renders the protein unable to exit the TGN. Bulky or charged substitutions at position 653 mimic the phenotype of the patient mutation. Molecular modeling and dynamic simulation suggest that the S653Y mutation induces local distortions within the transmembrane (TM) domain 1 and alter TM1 interaction with TM2. S653Y abolishes the trafficking-stimulating effects of a secondary mutation in the N-terminal apical targeting domain. This result indicates a role for TM1/TM2 in regulating conformations of cytosolic domains involved in ATP7B trafficking. Taken together, our experiments revealed an unexpected role for TM1/TM2 in copper-regulated trafficking of ATP7B and defined a unique class of WD mutants that are transport-competent but trafficking-defective. Understanding the precise consequences of WD-causing mutations will facilitate the development of advanced mutation-specific therapies.
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Continuous cast copper (O), BCR®, certified reference material, rod
Copper, sphere, 100pcs, diameter 3.0mm, precision sphere grade 200
Copper, tube, 1000mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 1.0mm, inside diameter 0.24mm, wall thickness 0.38mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Copper, tube, 1000mm, outside diameter 1.2mm, inside diameter 0.56mm, wall thickness 0.32mm, as drawn, 99.9%
Copper, tube, 100mm, outside diameter 1.75mm, inside diameter 0.33mm, wall thickness 0.71mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 12.7mm, inside diameter 10.26mm, wall thickness 1.22mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 12.7mm, as drawn, 99.99+%
Copper, tube, 1000mm, outside diameter 5.0mm, inside diameter 4mm, wall thickness 0.5mm, as drawn, 99.9%
Copper, tube, 1000mm, outside diameter 0.95mm, inside diameter 0.29mm, wall thickness 0.33mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 5m coil, thickness 0.05mm, coil width 300mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.075mm, half hard, 99.95+%
Copper, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.99+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.075mm, half hard, 99.95+%
Copper, tube, 500mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.125mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10m coil, thickness 0.125mm, hard, 99.95+%
Copper, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.99+%
Copper, tube, 500mm, outside diameter 2.5mm, inside diameter 1.9mm, wall thickness 0.3mm, as drawn, 99.9%