Merck
  • Protein adsorption to poly(ethylenimine)-modified Sepharose FF: III. Comparison between different proteins.
Protein adsorption to poly(ethylenimine)-modified Sepharose FF: III. Comparison between different proteins.
Journal of chromatography. A (2014-04-02)
Yan Hong, Na Liu, Wei Wei, Lin-Ling Yu, Guanghui Ma, Yan Sun
ABSTRACT
Previously, we studied bovine serum albumin (BSA) uptake to poly(ethylenimine) (PEI)-grafted Sepharose resins, and an ionic capacity (IC) range (600-740mmol/L) for steep increases of both protein capacity (qm) and effective pore diffusion coefficient (De) was found. In this work, seven PEI-grafted Sepharose FF resins at IC range of 270-1030mmol/L were synthesized to investigate the effect of protein properties on the adsorption and uptake kinetics using BSA and γ-globulin as two model proteins. For BSA, the change trends of qm and De values with IC were well consistent with the previous results. For γ-globulin, the qm values increased slowly till reaching a maximum value at IC=560mmol/L and then decreased rapidly at IC>560mol/L. The De values nearly kept unchanged at low ICs (IC<460mmol/L), and increased steeply at IC>460mmol/L till reaching a maximum at 680mmol/L (De/D0=0.48±0.01). After that increase, the De values for γ-globulin dropped quickly at IC>680mol/L, which was not observed for BSA. It is interesting to note that in the narrow IC range of 460-680mmol/L, the De values of γ-globulin increased dramatically for more than four folds. Moreover, it is notable that the IC range where the hopping of De values occurred for γ-globulin was earlier than that for BSA (460 vs. 560mmol/L). The earlier hopping of γ-globulin uptake rate was attributed to its larger size and less net charge, which facilitated the happenings of the "chain delivery" effect. The quick drops of both qm and De values for γ-globulin at IC>680mmol/L were considered due to its large size, which led to the significant decrease of its effective pore volume. The results indicate that both PEI layer and protein size played important roles in protein adsorption to PEI-grafted resins, and further prove the "chain delivery" effect did contributed significantly to the uptake rate hopping in the PEI-grafted resins. This work could also help the design and selection of resins based on protein characteristics and benefit optimization of practical chromatographic processes for therapeutic proteins with PEI-grafted anion exchangers.
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter
Sepharose 4B, Cytiva 17-0120-01, pack of 1 L
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sepharose 6B, Cytiva 17-0110-01, pack of 1 L
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, High EEO