Merck
  • Preclinical evaluation of marketed sodium channel blockers in a rat model of myotonia discloses promising antimyotonic drugs.
Preclinical evaluation of marketed sodium channel blockers in a rat model of myotonia discloses promising antimyotonic drugs.
Experimental neurology (2014-03-13)
Jean-François Desaphy, Roberta Carbonara, Teresa Costanza, Diana Conte Camerino
ABSTRACT
Although the sodium channel blocker mexiletine is considered the first-line drug in myotonia, some patients experiment adverse effects, while others do not gain any benefit. Other antimyotonic drugs are thus needed to offer mexiletine alternatives. In the present study, we used a previously-validated rat model of myotonia congenita to compare six marketed sodium channel blockers to mexiletine. Myotonia was induced in the rat by injection of anthracen-9-carboxylic acid, a muscle chloride channel blocker. The drugs were given orally and myotonia was evaluated by measuring the time of righting reflex. The drugs were also tested on sodium currents recorded in a cell line transfected with the human skeletal muscle sodium channel hNav1.4 using patch-clamp technique. In vivo, carbamazepine and propafenone showed antimyotonic activity at doses similar to mexiletine (ED50 close to 5mg/kg); flecainide and orphenadrine showed greater potency (ED50 near 1mg/kg); lubeluzole and riluzole were the more potent (ED50 near 0.1mg/kg). The antimyotonic activity of drugs in vivo was linearly correlated with their potency in blocking hNav1.4 channels in vitro. Deviation was observed for propafenone and carbamazepine, likely due to pharmacokinetics and multiple targets. The comparison of the antimyotonic dose calculated in rats with the current clinical dose in humans strongly suggests that all the tested drugs may be used safely for the treatment of human myotonia. Considering the limits of mexiletine tolerability and the occurrence of non-responders, this study proposes an arsenal of alternative drugs, which may prove useful to increase the quality of life of individuals suffering from non-dystrophic myotonia. Further clinical trials are warranted to confirm these results.
Product Description
Supelco
(±)-Flecainide solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Orphenadrine hydrochloride, ≥98.0% (AT)
Sigma-Aldrich
Propafenone hydrochloride
Sigma-Aldrich
Flecainide acetate salt
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Orphenadrine for peak identification, European Pharmacopoeia (EP) Reference Standard
Flecainide acetate, European Pharmacopoeia (EP) Reference Standard
Carbamazepine, European Pharmacopoeia (EP) Reference Standard
Orphenadrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Orphenadrine citrate, European Pharmacopoeia (EP) Reference Standard
USP
Carbamazepine, United States Pharmacopeia (USP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Riluzole, solid
Sigma-Aldrich
Carbamazepine, powder
Supelco
Carbamazepine, analytical standard
Propafenone hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Orphenadrine citrate salt, analytical standard
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Lubeluzole dihydrochloride, ≥98% (HPLC)