Merck
  • The accumulation and efflux of lead partly depend on ATP-dependent efflux pump-multidrug resistance protein 1 and glutathione in testis Sertoli cells.
The accumulation and efflux of lead partly depend on ATP-dependent efflux pump-multidrug resistance protein 1 and glutathione in testis Sertoli cells.
Toxicology letters (2014-03-07)
Shaoxin Huang, Jingping Ye, Jun Yu, Li Chen, Langhuan Zhou, Hong Wang, Zhen Li, Chunhong Wang
ABSTRACT
Since lead accumulation is toxic to cells, its excretion is crucial for organisms to survive the toxicity. In this study, mouse testis sertoli (TM4) and Mrp1 lower-expression TM4-sh cells were used to explore the lead accumulation characteristics, and the role of ATP-dependent efflux pump-multidrug resistance protein 1 (Mrp1) in lead excretion. TM4 cells possess Mrp-like transport activity. The expression levels of mrp1 mRNA and Mrp1 increased after lead treatments at first and then decreased. The maximum difference of relative mRNA expression reached 10 times. In the presence of lead acetate, the amount of cumulative lead in TM4-sh was much higher than that in TM4. After the treatment with lead acetate at 10-40 μM for 12h or 24h, the differences were about 2-8 times. After with the switch to lead-free medium, the cellular lead content in TM4-sh remains higher than that in TM4 cells at 1,3, 6, and 9h time points (P<0.01). Energy inhibitor sodium azide, Mrp inhibitors MK571 and glutathione (GSH) biosynthesis inhibitor BSO could block lead efflux from TM4 cells significantly. These results indicate that lead excretion may be mediated by Mrp1 and GSH in TM4 cells. Mrp1 could be one of the important intervention points for lead detoxification.
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 1.0mm, as rolled, 99.99+%
Lead, foil, 2m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, not light tested, 25x25mm, thickness 0.004mm, 99.99+%
Lead, microfoil, disks, 10mm, thinness 1.0μm, specific density 1047.8μg/cm2, permanent mylar 3.5μm support, 99.99+%
Lead, rod, 1000mm, diameter 3.2mm, 99.95%
Lead, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.999%
Lead, wire reel, 5m, diameter 3.0mm, as drawn, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.02mm, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.025mm, 99.99+%
Lead, wire reel, 1m, diameter 0.5mm, as drawn, 99.999%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Lead, foil, 25mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 2m coil, thickness 0.5mm, as rolled, 99.95%
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 10mm disks, thickness 0.02mm, 99.99+%
Lead, foil, 50x50mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 10mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.025mm, 99.99+%