Merck
  • Development of a novel ultrasound-assisted surfactant-enhanced emulsification microextraction method and its application to the analysis of eleven polycyclic aromatic hydrocarbons at trace levels in water.
Development of a novel ultrasound-assisted surfactant-enhanced emulsification microextraction method and its application to the analysis of eleven polycyclic aromatic hydrocarbons at trace levels in water.
Journal of chromatography. A (2011-03-23)
Jing Cheng, Guzalnur Matsadiq, Lu Liu, Yi-Wen Zhou, Gang Chen
ABSTRACT
A novel ultrasound-assisted surfactant-enhanced emulsification microextraction (UASEME) technique has been proposed by using low-density extraction solvents. In the proposed technique, Tween 80 and cyclohexane were injected into 5-mL glass test tubes with conical bottoms, containing 5.00 mL of a water sample that was located inside the ultrasonic bath. When the extraction process was finished, the glass test tube was sealed with a rubber plug and then placed upside down in a centrifuge. The finely dispersed droplets of cyclohexane collected at the conical bottom of test tube because the density of cyclohexane is less than of water, and the PAHs were concentrated in the cyclohexane. Next, 5 μL of the cyclohexane that collected at the conical bottom was removed using a 10-μL microsyringe and injected into high performance liquid chromatography coupled with fluorescence detection (HPLC-FLD) for analysis. The proposed method avoided the use of chlorinated solvents, which have been widely used as extraction solvents in a normal UASEME assay. Parameters that affected the extraction efficiency, such as the type and volume of the extraction solvent, the type and concentration of the surfactant, and the ultrasound emulsification time and salt addition, were investigated and optimised for the method. Under the optimum conditions, the enrichment factors ranged between 90 and 247. The limits of detection of the method were 0.6-62.5 ng L(-1). Good recoveries and repeatability of the method for the eleven PAHs were also obtained. The proposed UASEME technique has been demonstrated to be simple, practical and environmentally friendly for the determination of PAH residues in real water samples.
Product Description
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub