Merck
  • Pulsed-field gel electrophoresis typing of Escherichia coli strains from samples collected before and after pivmecillinam or placebo treatment of uncomplicated community-acquired urinary tract infection in women.
Pulsed-field gel electrophoresis typing of Escherichia coli strains from samples collected before and after pivmecillinam or placebo treatment of uncomplicated community-acquired urinary tract infection in women.
Journal of clinical microbiology (2006-05-05)
Karen Ejrnaes, Dorthe Sandvang, Bettina Lundgren, Sven Ferry, Stig Holm, Tor Monsen, Rolf Lundholm, Niels Frimodt-Moller
ABSTRACT
The primary infecting Escherichia coli strains from 156 women with community-acquired uncomplicated urinary tract infection (UTI) randomized to pivmecillinam or placebo and the E. coli strains causing UTI at two follow-up visits were typed using pulsed-field gel electrophoresis (PFGE). In the pivmecillinam treatment group PFGE showed that among patients having a negative urine culture at the first follow-up 77% (46/60) had a relapse with the primary infecting E. coli strain and 23% (14/60) had reinfection with a new E. coli strain at the second follow-up. Among patients having E. coli at the first follow-up PFGE showed that 80% (32/40) had persistence with the primary infecting E. coli strain, 15% (6/40) had reinfection with a new E. coli strain, and 5% (2/40) had different E. coli strains at the two follow-up visits (one had reinfection followed by relapse, and the other had persistence followed by reinfection). In the placebo group the majority had E. coli at the first follow-up. PFGE showed that among these patients 96% (50/52) had persistence with the primary infecting E. coli strain and 4% (2/50) had different E. coli strains at the two follow-up visits (both had persistence followed by reinfection). The finding that the majority of UTIs at follow-up are caused by the primary infecting E. coli strain supports the theory of a vaginal and rectal reservoir but could also support the recent discovery that E. coli strains are able to persist in the bladder epithelium despite appropriate antibiotic treatment, constituting a reservoir for recurrent UTI.
Sigma-Aldrich
Pivmecillinam, ≥98% (HPLC)