Merck
Tailoring the mesoporous texture of graphitic carbon nitride.
Journal of nanoscience and nanotechnology (2013-11-20)
Jae-Hun Yang, Gain Kim, Kazunari Domen, Jin-Ho Choy
ABSTRACT
Recently, graphitic carbon nitride (g-C3N4) materials have received a great attention from many researchers due to their various roles as a visible light harvesting photocatalyst, metal-free catalyst, reactive template, nitrogen source of nitridation reaction, etc. g-C3N4 could be prepared by temperature-induced polymerization of cyanamide or melamine. In this study, we report a preparation of mesoporous graphitic carbon nitrides with tailored porous texture including pore size, and specific surface area from cyanamide and colloidal silica nanoparticles (Ludox). At first, cyanamide-silica nanocomposites were prepared by mixing colloidal silica with different size in the range of 7-22 nm and cyanamide, followed by evaporating the solvent in the resulting mixture. Mesoporous g-C3N4 samples were prepared by calcining cyanamide-silica nanocomposite at 550 degrees C for 4 hrs and removing the silica nanoparticles by using ammonium hydrogen fluoride. The formation of g-C3N4 was confirmed by the sharp (002) peak (d = 3.25 A) of graphitic interlayer stacking, and the broad (100) peak (d = 6.86 A) of in-plane repeating unit in the X-ray diffraction patterns. According to N2 adsorption-desorption analysis, the pore size of mesoporous carbon nitrides was similar to the size of colloidal silica used as hard template (7-22 nm). The specific surface area of mesoporous g-C3N4 could be tailored in the range of 189 m2/g-288 m2/g.
Sigma-Aldrich
Graphite, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Sigma-Aldrich
Graphite, flakes
Sigma-Aldrich
Graphite, powder, <20 μm, synthetic
Sigma-Aldrich
Graphite, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphite, powder, <45 μm, ≥99.99% trace metals basis