Merck
Denture labeling: A new approach.
Contemporary clinical dentistry (2011-10-01)
Pardeep K Bansal, Akshey Sharma, Rajesh Bhanot
ABSTRACT
The need for denture labeling is important for forensic and social reasons in case patients need to be identified individually. The importance of denture marking has long been acknowledged by the dental profession. Over the years, various denture marking systems have been reported in the literature, but none till date fulfills all the prescribed ADA specifications. A simple, easy, inexpensive procedure for marking accurate identification marks on dentures with a lead foil is described here. The label caring the patient information is incorporated in the acrylic resin during the denture processing.
Product Description
Lead, foil, light tested, 50x50mm, thickness 0.01mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Lead, foil, 6mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.015mm, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.03mm, 99.99+%
Lead, foil, 10mm disks, thickness 0.04mm, 99.99+%
Lead, foil, 5m coil, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 50mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.009mm, 99.99+%
Lead, foil, 50x50mm, thickness 1.5mm, as rolled, 99.99+%
Lead, foil, light tested, 100x100mm, thickness 0.04mm, 99.99+%
Lead, foil, 0.5m coil, thickness 0.016mm, as rolled, 96% (sn 1.5%, sb 2.5%)
Lead, foil, 1m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 4mm disks, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 100x100mm, thickness 0.15mm, as rolled, 99.95%
Lead, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Lead, foil, 8mm disks, thickness 0.01mm, 99.99+%
Lead, foil, 25mm disks, thickness 0.05mm, 99.99+%
Lead, foil, 50x50mm, thickness 6.0mm, as rolled, 99.99+%
Lead, foil, 6mm disks, thickness 0.03mm, 99.99+%
Lead, foil, 50mm disks, thickness 0.25mm, as rolled, 99.95%
Lead, foil, 15mm disks, thickness 0.015mm, 99.99+%
Lead, foil, 5m coil, thickness 0.1mm, as rolled, 99.95%
Lead, foil, 25mm disks, thickness 0.025mm, 99.99+%
Lead, foil, 15mm disks, thickness 0.1mm, as rolled, 99.95%