Merck
  • Time-resolved EUV photoelectron spectroscopy of dissociating I2 by laser harmonics at 80 nm.
Time-resolved EUV photoelectron spectroscopy of dissociating I2 by laser harmonics at 80 nm.
Optics express (2011-06-07)
Mizuho Fushitani, Akitaka Matsuda, Akiyoshi Hishikawa
ABSTRACT
Generation of single-order laser harmonics in extreme ultraviolet (EUV) and the application to the time-resolved photoelectron spectroscopy of I(2) are demonstrated. The EUV pulses at 80 nm were generated from Kr as the 5th order harmonics of intense 400 nm laser pulses and then separated from other harmonic orders by a thin indium foil. The pump-probe photoelectron spectroscopy of I(2) in the B (3)Π(0(u)(+)) and B" (1)Π(1(u)) states excited by visible laser pulses at 490 nm showed a rapid increase in the yield of atomic iodine (~400 fs), reflecting the dissociation dynamics evolving simultaneously in the two excited states.
Product Description
Indium, foil, 4mm disks, thickness 0.01mm, 99.8%
Indium, foil, 15mm disks, thickness 0.125mm, as rolled, 100%
Indium, foil, 4mm disks, thickness 0.25mm, as rolled, 100%
Indium, foil, 50x50mm, thickness 0.75mm, as rolled, 99.999%
Indium, foil, not light tested, 150x150mm, thickness 0.075mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.02mm, 99.8%
Indium, foil, not light tested, 75x300mm, thickness 0.05mm, as rolled, 100%
Indium, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 100%
Indium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.8%
Indium, foil, 100x100mm, thickness 0.20mm, as rolled, 99.999%
Indium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Indium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.999%
Indium, foil, 25x25mm, thickness 2.0mm, as rolled, 99.999%
Indium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.999%
Indium, foil, 150x150mm, thickness 0.20mm, as rolled, 99.999%
Indium, foil, 25x25mm, thickness 3.0mm, as rolled, 99.999%
Indium, foil, 100x100mm, thickness 0.75mm, as rolled, 99.999%
Indium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.999%
Indium, foil, 50x100mm, thickness 0.1mm, as rolled, 100%
Indium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%
Indium, foil, 150x150mm, thickness 0.75mm, as rolled, 99.999%
Indium, foil, 10mm disks, thickness 0.04mm, 99.8%
Indium, foil, 50x50mm, thickness 0.20mm, as rolled, 99.999%
Indium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.999%
Indium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.999%
Indium, foil, 8mm disks, thickness 0.03mm, 99.8%
Indium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%
Indium, foil, 6mm disks, thickness 0.25mm, as rolled, 100%
Indium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.8%
Indium, foil, 8mm disks, thickness 0.04mm, 99.8%