Merck
Fused-silica focusing lens for deep UV laser processing.
Applied optics (1992-08-01)
T Y Hung, C S Su
ABSTRACT
A simple four-element fused-silica lens is presented that has a focal length of 31.2 mm and a relative aperture of f/1 for use as a focusing lens for deep UV laser processing. The curvature of the lens is designed with a practical design method to match the curvature of existing optical tools. By applying an 8.5-mm beam diameter Nd:YAG laser at a wavelength of 266 nm through the lens, we can easily achieve the percussion drilled into samples of 25-microm-thick tantalum foil with a hole diameter of <6 microm. When the lens and/or the sample is tilted, the shape of the drilled hole is in agreement with the Gaussian beam spot simulation.
Product Description
Tantalum, foil, light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.015mm, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0015mm, 99.9%
Tantalum, foil, light tested, 150x300mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.01mm, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.003mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.02mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.003mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0015mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.009mm, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.006mm, 99.9%
Tantalum, foil, not light tested, 82x100mm, thickness 0.006mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0005mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.01mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.001mm, 99.9%
Tantalum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Tantalum, foil, 100x100mm, thickness 0.100mm, annealed, 99.9%
Tantalum, foil, 50x50mm, thickness 5.0mm, annealed, 99.9%
Tantalum, foil, 0.5m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Tantalum, foil, 50x50mm, thickness 0.125mm, annealed, 99.9%
Tantalum, foil, 100x100mm, thickness 1.5mm, annealed, 99.9%
Tantalum, foil, 10m coil, thickness 0.025mm, coil width 0.75mm, annealed, 99.9%
Tantalum, foil, 50x50mm, thickness 0.5mm, annealed, 99.9%
Tantalum, foil, 0.2m coil, thickness 0.03mm, coil width 1.2mm, annealed, 99.9%
Tantalum, foil, 0.2m coil, thickness 0.1mm, coil width .62mm, as rolled, 99.9%
Tantalum, foil, 10m coil, thickness 0.03mm, coil width 0.75mm, annealed, 99.9%