Merck
  • Characterization of aroma-active compounds in raw and cooked pine-mushrooms (Tricholoma matsutake Sing.).
Characterization of aroma-active compounds in raw and cooked pine-mushrooms (Tricholoma matsutake Sing.).
Journal of agricultural and food chemistry (2006-08-17)
In Hee Cho, Se Young Kim, Hyung-Kyoon Choi, Young-Suk Kim
ABSTRACT
The characteristic aroma-active compounds in raw and cooked pine-mushrooms (Tricholoma matsutake Sing.) were investigated by gas chromatography-olfactometry using aroma extract dilution analysis. 1-Octen-3-one (mushroom-like) was the major aroma-active compound in raw pine-mushrooms; this compound had the highest flavor dilution factor, followed by ethyl 2-methylbutyrate (floral and sweet), linalool (citrus-like), methional (boiled potato-like), 3-octanol (mushroom-like and buttery), 1-octen-3-ol (mushroom-like), (E)-2-octen-1-ol (mushroom-like), and 3-octanone (mushroom-like and buttery). By contrast, methional, 2-acetylthiazole (roasted), an unknown compound (chocolate-like), 3-hydroxy-2-butanone (buttery), and phenylacetaldehyde (floral and sweet), which could be formed by diverse thermal reactions during the cooking process, together with C8 compounds, were identified as the major aroma-active compounds in cooked pine-mushrooms.
Sigma-Aldrich
2-Acetylthiazole, ≥99%, FG
Sigma-Aldrich
1-Octen-3-one, 50 wt. % in 1-octen-3-ol, stabilized, FG
Sigma-Aldrich
Phenylacetaldehyde, ≥90%
Sigma-Aldrich
Phenylacetaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
1-Octen-3-one, 96%