Merck
  • Renal effects of selective cyclooxygenase-2 inhibition in normotensive salt-depleted subjects.
Renal effects of selective cyclooxygenase-2 inhibition in normotensive salt-depleted subjects.
Clinical pharmacology and therapeutics (1999-08-03)
J Rossat, M Maillard, J Nussberger, H R Brunner, M Burnier
ABSTRACT
To compare the renal hemodynamic and tubular effects of celecoxib, a selective inhibitor of cyclooxygenase-2 (COX-2) to those of naproxen, a nonselective inhibitor of cyclooxygenases in salt-depleted subjects. Forty subjects were randomized into four parallel groups to receive 200 mg celecoxib twice a day, 400 mg celecoxib twice a day, 500 mg naproxen twice a day, or a placebo for 7 days according to a double-blind study design. Blood pressure, renal hemodynamics, and urinary water and electrolyte excretion were measured before and for 3 hours after drug intake on days 1 and 7. Celecoxib had no effect on systemic blood pressure, but short-term transient decreases in renal blood flow and glomerular filtration rate were found with the highest dose of 400 mg on day 1. On the first day, both celecoxib and naproxen decreased urine output (P < .05) and sodium, lithium, and potassium excretion (P < .01). On day 7, similar effects on water and sodium excretion were observed. During repeated administration, a significant sodium retention occurred during the first 3 days. In salt-depleted subjects, selective inhibition of COX-2 causes sodium and potassium retention. This suggests that an increased selectivity for COX-2 does not spare the kidney, at least during salt depletion.
Product Description
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard