Merck
  • A liquid chromatography and tandem mass spectrometry method for the determination of potential biomarkers of cardiovascular disease.
A liquid chromatography and tandem mass spectrometry method for the determination of potential biomarkers of cardiovascular disease.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-02-16)
Sylwia Magiera, Irena Baranowska, Jacek Kusa, Jacek Baranowski
ABSTRACT
A simple, accurate and sensitive liquid chromatography tandem mass spectrometry (LC-MS/MS) method was developed and validated for the quantitation of α-ketoglutaric acid (α-KG), L-carnitine (L-CAR) and acetyl-L-carnitine (acetyl-L-CAR) in human urine as potential biomarkers of cardiovascular disease. The separation was performed using an isocratic elution of 0.1% formic acid in water and acetonitrile (97:3, v/v) on an Acclaim 120 C8 column (150 mm × 4.6 mm, 3.0 μm). The flow rate of the mobile phase was 1.2 mL/min and the total assay run time was 3 min. Detection was performed on a triple-quadrupole mass spectrometer in selected reaction monitoring (SRM) mode via an electrospray ionization (ESI) source in positive and negative ion modes. This method covered a linearity range of 0.1-500 ng/mL for L-CAR and acetyl-L-CAR and 1-1000 ng/mL for α-KG with lower limits of quantification (LLOQ) of 0.08 ng/mL for L-CAR, 0.04 ng/mL for acetyl-L-CAR and 0.8 ng/mL for α-KG. The intra-day and inter-day precision and accuracy of the quality control samples exhibited relative standard deviations of less than 5.54% and relative error values from -5.95% to 3.11%. Analyte stability was evaluated under various sample preparation, analysis and storage conditions and varied from -9.89% to -0.47%. A two-step solid-phase extraction (SPE) procedure using silica gel and quaternary amine cartridges was used for urine sample cleanup. The average recoveries for all analyzed compounds were better than 86.64% at three concentrations. The method was successfully applied for the quantitation of α-KG, L-CAR and acetyl-L-CAR in human urine samples.
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
Levocarnitine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)
Sigma-Aldrich
L-Carnitine inner salt, synthetic, ≥98%