Merck
  • Development of whispering gallery mode polymeric micro-optical electric field sensors.
Development of whispering gallery mode polymeric micro-optical electric field sensors.
Journal of visualized experiments : JoVE (2013-02-15)
Tindaro Ioppolo, Volkan Ötügen, Ulas Ayaz
ABSTRACT
Optical modes of dielectric micro-cavities have received significant attention in recent years for their potential in a broad range of applications. The optical modes are frequently referred to as "whispering gallery modes" (WGM) or "morphology dependent resonances" (MDR) and exhibit high optical quality factors. Some proposed applications of micro-cavity optical resonators are in spectroscopy, micro-cavity laser technology, optical communications as well as sensor technology. The WGM-based sensor applications include those in biology, trace gas detection, and impurity detection in liquids. Mechanical sensors based on microsphere resonators have also been proposed, including those for force, pressure, acceleration and wall shear stress. In the present, we demonstrate a WGM-based electric field sensor, which builds on our previous studies. A candidate application of this sensor is in the detection of neuronal action potential. The electric field sensor is based on polymeric multi-layered dielectric microspheres. The external electric field induces surface and body forces on the spheres (electrostriction effect) leading to elastic deformation. This change in the morphology of the spheres, leads to shifts in the WGM. The electric field-induced WGM shifts are interrogated by exciting the optical modes of the spheres by laser light. Light from a distributed feedback (DFB) laser (nominal wavelength of ~ 1.3 μm) is side-coupled into the microspheres using a tapered section of a single mode optical fiber. The base material of the spheres is polydimethylsiloxane (PDMS). Three microsphere geometries are used: (1) PDMS sphere with a 60:1 volumetric ratio of base-to-curing agent mixture, (2) multi layer sphere with 60:1 PDMS core, in order to increase the dielectric constant of the sphere, a middle layer of 60:1 PDMS that is mixed with varying amounts (2% to 10% by volume) of barium titanate and an outer layer of 60:1 PDMS and (3) solid silica sphere coated with a thin layer of uncured PDMS base. In each type of sensor, laser light from the tapered fiber is coupled into the outermost layer that provides high optical quality factor WGM (Q ~ 10(6)). The microspheres are poled for several hours at electric fields of ~ 1 MV/m to increase their sensitivity to electric field.
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)