Merck
  • Influence of composition on the performance of sintered Cu(In,Ga)Se2 nanocrystal thin-film photovoltaic devices.
Influence of composition on the performance of sintered Cu(In,Ga)Se2 nanocrystal thin-film photovoltaic devices.
ChemSusChem (2013-02-13)
Vahid A Akhavan, Taylor B Harvey, C Jackson Stolle, David P Ostrowski, Micah S Glaz, Brian W Goodfellow, Matthew G Panthani, Dariya K Reid, David A Vanden Bout, Brian A Korgel
ABSTRACT
Thin-film photovoltaic devices (PVs) were prepared by selenization using oleylamine-capped Cu(In,Ga)Se2 (CIGS) nanocrystals sintered at a high temperature (>500 °C) under Se vapor. The device performance varied significantly with [Ga]/[In+Ga] content in the nanocrystals. The highest power conversion efficiency (PCE) observed in the devices studied was 5.1 % under air mass 1.5 global (AM 1.5 G) illumination, obtained with [Ga]/[In+Ga]=0.32. The variation in PCE with composition is partly a result of bandgap tuning and optimization, but the main influence of nanocrystal composition appeared to be on the quality of the sintered films. The [Cu]/[In+Ga] content was found to be strongly influenced by the [Ga]/[In+Ga] concentration, which appears to be correlated with the morphology of the sintered film. For this reason, only small changes in the [Ga]/[In+Ga] content resulted in significant variations in device efficiency.
Sigma-Aldrich
Indium, bars, 99.95%
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 2.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, beads, diam. 2-5 mm, 99.999% trace metals basis
Sigma-Aldrich
Indium, wire, diam. 0.45-0.55 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, powder, 99.99% trace metals basis
Sigma-Aldrich
Indium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Indium, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Indium, pieces, 99.99% trace metals basis
Sigma-Aldrich
Indium, rod, diam. 6 mm, ≥99.999% trace metals basis