Merck
  • Dichloro-dihydro-fluorescein diacetate (DCFH-DA) assay: a quantitative method for oxidative stress assessment of nanoparticle-treated cells.
Dichloro-dihydro-fluorescein diacetate (DCFH-DA) assay: a quantitative method for oxidative stress assessment of nanoparticle-treated cells.
Toxicology in vitro : an international journal published in association with BIBRA (2013-01-30)
A Aranda, L Sequedo, L Tolosa, G Quintas, E Burello, J V Castell, L Gombau
ABSTRACT
No consensus exists on how to address possible toxicity of nanomaterials as they interfere with most in vitro screening tests based on colorimetric and fluorimetric probes such as the dichloro-dihydro-fluorescein diacetate (DCFH-DA) assay for detection of oxidative species. In the present research, nanomaterial interaction with DCFH-DA was studied in relation to its nature and/or assay conditions (cell-based and time exposure) by incubating Rhodamine (Rhd)-labeled 25nm and 50nm silica (SiO2), naked and oleic acid coated magnetite, (Fe3O4) and maghemite (Fe2O3) iron oxide, titanium dioxide (TiO2) and poly(ethylene oxide)-poly(lactide/glycolide) acid (PLGA-PEO) nanoparticles (NPs) with metabolically active rat hepatocytes for 4 and 24-h periods. Data indicated that nanoparticle uptake correlated with quenching of dye fluorescence emission. In spite of their masking effect, the oxidative potential of NPs could be detected at a limited threshold concentration when exposed for periods of time longer than those frequently used for this test. However, changes in the experimental conditions did not systematically result in free radical formation for all nanomaterials tested. Overall data indicate that despite the quenching effect of nanoparticles on DCFH-DA assay, it can be considered as a useful tool for quantitative measurement of NPs-induced oxidative stress by minor modifications of standardized protocols.
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in toluene
Sigma-Aldrich
Iron(II,III) oxide, nanopowder, 50-100 nm particle size (SEM), 97% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, 99.99% trace metals basis
Sigma-Aldrich
Iron(II,III) oxide, powder, <5 μm, 95%
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 10 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 20 nm avg. part. size, 5 mg/mL in H2O
Sigma-Aldrich
Iron oxide(II,III), magnetic nanoparticles solution, 5 nm avg. part. size, 5 mg/mL in H2O