Merck
A DFT + U study of (Rh, Nb)-codoped rutile TiO2.
Journal of physics. Condensed matter : an Institute of Physics journal (2013-01-25)
Kulbir Kaur Ghuman, Chandra Veer Singh
ABSTRACT
A systematic study of electronic structure and band gap states is conducted to analyze the monodoping and charge compensated codoping of rutile TiO(2) with Rh and Nb, using the DFT + U approach. Doping of rutile TiO(2) with Rh atoms induces hybridized O 2p and Rh 4d band gap states leading to a red shift of the optical absorption edge, consistent with previous experimental studies. Since Rh monodoping may induce recombination centers, charge compensated codoping with Rh and Nb is also explored. This codoping induces an electron transfer from Nb induced states to Rh 4d states, which suppresses the formation of Rh(4+), thereby leading to a reduction in recombination centers and to the formation of more stable Rh(3+). A combination of band gap reduction by 0.5 eV and the elimination of band gap states that account for recombination centers makes (Rh, Nb)-codoped TiO(2) a more efficient and stable photocatalyst.
Product Description
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire