Merck
  • Nb-Nb interactions define the charge density wave structure of 2H-NbSe2.
Nb-Nb interactions define the charge density wave structure of 2H-NbSe2.
Journal of the American Chemical Society (2013-01-23)
Christos D Malliakas, Mercouri G Kanatzidis
ABSTRACT
2H-NbSe(2) is a canonical Charge-Density-Wave (CDW) layered material the structural details of which remained elusive. We report the detailed structure of 2H-NbSe(2) below the CDW transition using a (3 + 2)-dimensional crystallographic approach on single crystal X-ray diffraction data collected at 15 K. Intensities of main reflections as well as CDW satellites of first order were measured. Quantitative information about the magnitude of the structural distortions and clustering of Nb atoms were extracted from the refined model. The Nb-Nb distances were found to distort between 3.4102(8) and 3.4928(8) Å in the CDW phase from the average undistorted distance of 3.4583(4) Å.
Product Description
Niobium, IRMM®, certified reference material, 0.5 mm wire
Niobium, IRMM®, certified reference material, 0.02 mm foil
Sigma-Aldrich
Niobium, powder, <45 μm, 99.8% trace metals basis
Sigma-Aldrich
Niobium, foil, thickness 0.25 mm, 99.8% trace metals basis
Niobium, IRMM®, certified reference material, 0.02 mm foil
Niobium, IRMM®, certified reference material, 0.5 mm wire