Merck
  • The use of agarose microwells for scalable embryoid body formation and cardiac differentiation of human and murine pluripotent stem cells.
The use of agarose microwells for scalable embryoid body formation and cardiac differentiation of human and murine pluripotent stem cells.
Biomaterials (2013-01-22)
Julia Dahlmann, George Kensah, Henning Kempf, David Skvorc, Anke Gawol, David A Elliott, Gerald Dräger, Robert Zweigerdt, Ulrich Martin, Ina Gruh
ABSTRACT
In most pluripotent stem cell differentiation protocols the formation of embryoid bodies (EBs) is an important step. Here we describe a rapid, straightforward soft lithography approach for the preparation of hydrophilic silicon masters from different templates and the subsequent production of patterned agarose-DMEM microwell surfaces for scalable well standardized stem cell aggregation and EB formation. The non-adhesive agarose microwell plates represent an accurate replication of the templates' topography and were used for aggregation of murine induced pluripotent stem cells (iPSCs) and human embryonic stem cells (ESCs). Direct microscopic assessment by time-lapse analysis demonstrated rapid formation of uniformly shaped EBs from murine iPSCs with similar or even more consistent results concerning size distribution and harvesting efficiency compared to the commonly used but time-consuming hanging drop technique. For human ESCs, homogenous aggregates were obtained after single cell inoculation on agarose microwells with efficient differentiation into the cardiac lineage using state-of-the-art protocols for directed differentiation via small molecules. With this soft lithography-based strategy, sufficient and reproducible numbers of stem cell-derived cardiomyocytes necessary for tissue engineering purposes can be realized in a highly controllable manner. Moreover, it might be useful for different cell types in any application that requires scalable and highly standardized aggregation.
Sigma-Aldrich
Agarose, Medium EEO, for molecular biology
Sigma-Aldrich
Agarose, Wide range, for molecular biology
Sigma-Aldrich
Agarose, for molecular biology
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, Wide range/Standard 3:1
Sigma-Aldrich
Agarose, Type I, low EEO
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature
Sigma-Aldrich
Agarose, For pulsed field electrophoresis running gel
Sigma-Aldrich
Agarose, Low EEO, for Immunoelectrophoresis
Sigma-Aldrich
Agarose, Special High EEO
Sigma-Aldrich
Agarose, BioReagent, for molecular biology, low EEO
Sigma-Aldrich
Agarose, High Gelling Temperature
Sigma-Aldrich
Agarose, Ultra-low Gelling Temperature, molecular biology grade
Sigma-Aldrich
Agarose, Type II-A, Medium EEO
Sigma-Aldrich
Sepharose 6B, 6% Beaded Agarose, 45-165 μm (wet), fractionation range 10,000-1,000,000 Da (dextrans)
Sigma-Aldrich
Agarose, Type IV, Special High EEO
Sigma-Aldrich
Agarose, Type I-A, low EEO
Sigma-Aldrich
Agarose, Low EEO
Sigma-Aldrich
Agarose, High EEO, for molecular biology
Sigma-Aldrich
Sepharose 4B, 45-165 μm bead diameter