Merck
  • PEGylation of vesicular stomatitis virus extends virus persistence in blood circulation of passively immunized mice.
PEGylation of vesicular stomatitis virus extends virus persistence in blood circulation of passively immunized mice.
Journal of virology (2013-01-18)
Mulu Z Tesfay, Amber C Kirk, Elizabeth M Hadac, Guy E Griesmann, Mark J Federspiel, Glen N Barber, Stephen M Henry, Kah-Whye Peng, Stephen J Russell
ABSTRACT
We are developing oncolytic vesicular stomatitis viruses (VSVs) for systemic treatment of multiple myeloma, an incurable malignancy of antibody-secreting plasma cells that are specifically localized in the bone marrow. One of the presumed advantages for using VSV as an oncolytic virus is that human infections are rare and preexisting anti-VSV immunity is typically lacking in cancer patients, which is very important for clinical success. However, our studies show that nonimmune human and mouse serum can neutralize clinical-grade VSV, reducing the titer by up to 4 log units in 60 min. In addition, we show that neutralizing anti-VSV antibodies negate the antitumor efficacy of VSV, a concern for repeat VSV administration. We have investigated the potential use of covalent modification of VSV with polyethylene glycol (PEG) or a function-spacer-lipid (FSL)-PEG construct to inhibit serum neutralization and to limit hepatosplenic sequestration of systemically delivered VSV. We report that in mice passively immunized with neutralizing anti-VSV antibodies, PEGylation of VSV improved the persistence of VSV in the blood circulation, maintaining a more than 1-log-unit increase in VSV genome copies for up to 1 h compared to the genome copy numbers for the non-PEGylated virus, which was mostly cleared within 10 min after intravenous injection. We are currently investigating if this increase in PEGylated VSV circulating half-life can translate to increased virus delivery and better efficacy in mouse models of multiple myeloma.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Pricing and availability is not currently available.