Merck
  • Losartan added to β-blockade therapy for aortic root dilation in Marfan syndrome: a randomized, open-label pilot study.
Losartan added to β-blockade therapy for aortic root dilation in Marfan syndrome: a randomized, open-label pilot study.
Mayo Clinic proceedings (2013-01-17)
Hsin-Hui Chiu, Mei-Hwan Wu, Jou-Kou Wang, Chun-Wei Lu, Shuenn-Nan Chiu, Chun-An Chen, Ming-Tai Lin, Fu-Chang Hu
ABSTRACT
To assess the tolerability and efficacy of the investigational use of the angiotensin II receptor blocker losartan added to β-blockade (BB) to prevent progressive aortic root dilation in patients with Marfan syndrome (MFS). Between May 1, 2007, and September 31, 2011, 28 patients with MFS (11 males [39%]; mean ± SD age, 13.1±6.3 years) with recognized aortic root dilation (z score >2.0) and receiving BB (atenolol or propranolol) treatment were enrolled. They were randomized to receive BB (BB: 13 patients) or β-blockade and losartan (BB-L: 15 patients) for 35 months. In the BB-L group, aortic root dilation was reduced with treatment, and the annual dilation rate of the aortic root was significantly lower than that of the BB group (0.10 mm/yr vs 0.89 mm/yr; P=.02). The absolute aortic diameters at the sinus of Valsalva, annulus, and sinotubular junction showed similar trends, with a reduced rate of dilation in the BB-L group (P=.02, P=.03, and P=.03, respectively). Five patients (33%) treated with BB-L were noted to have a reduced aortic root diameter. However, the differences between the groups regarding changes in aortic stiffness and cross-sectional compliance were not statistically significant. This randomized, open-label, active controlled trial mostly based on a pediatric population demonstrated for the first time that losartan add-on BB therapy is safe and provides more effective protection to slow the progression of aortic root dilation than does BB treatment alone in patients with MFS. clinicaltrials.gov Identifier: NCT00651235.
Product Description
Supelco
Atenolol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder