Merck
  • Au(III)-CTAB reduction by ascorbic acid: preparation and characterization of gold nanoparticles.
Au(III)-CTAB reduction by ascorbic acid: preparation and characterization of gold nanoparticles.
Colloids and surfaces. B, Biointerfaces (2013-01-10)
Zaheer Khan, Taruna Singh, Javed Ijaz Hussain, Athar Adil Hashmi
ABSTRACT
Upon addition of tetrachloroauric(III) (HAuCl(4)) to a solution of cetyltrimethylammonium bromide (CTAB), a perfect transparent yellow colored and yellowish-precipitate appears within the time of mixing, indicating the formation of a complex between HAuCl(4) and CTAB. Morphology of gold nanoparticles in situ via chemical reduction of title reaction has been determined by using conventional techniques. The position and shape of the surface plasmon resonance (SRP) band strongly depends on the [ascorbic acid], [HAuCl(4)], [CTAB] and mixing ratio of the reactants. Sub-, post- and dilution-micellar effects are accountable to the aggregation and/or adsorption of gold nanoparticles onto the surface of CTAB. The morphology of particles was altered by changing the [reactants] as well as [CTAB]. A mechanism of nanoparticle formation has been proposed comprising different steps of particle growth.
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%