Merck
  • Curcumin loaded polymeric micelles inhibit breast tumor growth and spontaneous pulmonary metastasis.
Curcumin loaded polymeric micelles inhibit breast tumor growth and spontaneous pulmonary metastasis.
International journal of pharmaceutics (2013-01-05)
Lei Liu, Lu Sun, Qinjie Wu, Wenhao Guo, Ling Li, YiShan Chen, Yuchen Li, Changyang Gong, Zhiyong Qian, Yuquan Wei
ABSTRACT
This work aims to develop curcumin (Cur) loaded biodegradable self-assembled polymeric micelles (Cur-M) to overcome poor water solubility of Cur and to meet the requirement of intravenous administration. Cur-M were prepared by solid dispersion method, which was simple and easy to scale up. Cur-M had a small particle size of 28.2 ± 1.8 nm and polydisperse index (PDI) of 0.136 ± 0.050, and drug loading and encapsulation efficiency of Cur-M were 14.84 ± 0.11% and 98.91 ± 0.70%, respectively. Besides, in vitro release profile showed a significant difference between rapid release of free Cur and much slower and sustained release of Cur-M. Cytotoxicity study showed that the encapsulated Cur remained its potent anti-tumor effect. Furthermore, Cur-M were more effective in inhibiting tumor growth and spontaneous pulmonary metastasis in subcutaneous 4T1 breast tumor model, and prolonged survival of tumor-bearing mice. In addition, immunofluorescent and immunohistochemical studies also showed that tumors of Cur-M-treated mice had more apoptosis cells, fewer microvessels, and fewer proliferation-positive cells. In conclusion, polymeric micelles encapsulating Cur were developed with enhanced anti-tumor and anti-metastasis activity on breast tumor, and Cur-M is excellent water-based formulation of Cur which may serve as a candidate for breast cancer therapy.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Pricing and availability is not currently available.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Pricing and availability is not currently available.