Merck
  • The mechanism of carvacrol-evoked [Ca2+]i rises and non-Ca2+-triggered cell death in OC2 human oral cancer cells.
The mechanism of carvacrol-evoked [Ca2+]i rises and non-Ca2+-triggered cell death in OC2 human oral cancer cells.
Toxicology (2012-11-14)
Wei-Zhe Liang, Chiang-Ting Chou, Ti Lu, Chao-Chuan Chi, Li-Ling Tseng, Chih-Chuan Pan, Ko-Long Lin, Chun-Chi Kuo, Chung-Ren Jan
ABSTRACT
Carvacrol is one of the main substances of essential oil which triggers intracellular Ca(2+) mobilization and causes cytotoxicity in diverse cell models. However, the mechanism of carvacrol-induced Ca(2+) movement and cytotoxicity is not fully understood. This study examined the effect of carvacrol on cytosolic free Ca(2+) concentrations ([Ca(2+)](i)), cell viability and apoptosis in OC2 human oral cancer cells. Carvacrol induced a [Ca(2+)](i) rise and the signal was reduced by removal of extracellular Ca(2+). Carvacrol-induced Ca(2+) entry was not altered by store-operated Ca(2+) channel inhibitors and protein kinase C (PKC) activator, but was inhibited by a PKC inhibitor. In Ca(2+) -free medium, treatment with the endoplasmic reticulum Ca(2+) pump inhibitor thapsigargin (TG) or 2,5-di-tert-butylhydroquinone (BHQ) inhibited carvacrol-induced [Ca(2+)](i) rise. Conversely, incubation with carvacrol inhibited TG or BHQ-induced [Ca(2+)](i) rise. Inhibition of phospholipase C (PLC) with U73122 abolished carvacrol-induced [Ca(2+)](i) rise. Carvacrol decreased cell viability, which was not reversed when cytosolic Ca(2+) was chelated with BAPTA-AM (1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid-acetoxymethyl ester). Carvacrol-induced apoptosis and activation of reactive oxygen species (ROS) and caspase-3. Together, carvacrol induced a [Ca(2+)](i) rise by inducing PLC-dependent Ca(2+) release from the endoplasmic reticulum and Ca(2+) entry via PKC-sensitive, non store-operated Ca(2+) channels. Carvacrol-induced ROS- and caspase-3-associated apoptosis.
Sigma-Aldrich
Carvacrol, natural, 99%, FG
Sigma-Aldrich
Carvacrol, ≥98%, FCC, FG
Sigma-Aldrich
Carvacrol, 98%
Supelco
Carvacrol, analytical standard