Merck
  • Acid-labile mPEG-vinyl ether-1,2-dioleylglycerol lipids with tunable pH sensitivity: synthesis and structural effects on hydrolysis rates, DOPE liposome release performance, and pharmacokinetics.
Acid-labile mPEG-vinyl ether-1,2-dioleylglycerol lipids with tunable pH sensitivity: synthesis and structural effects on hydrolysis rates, DOPE liposome release performance, and pharmacokinetics.
Molecular pharmaceutics (2012-10-04)
Junhwa Shin, Pochi Shum, Jessica Grey, Shin-ichi Fujiwara, Guarov S Malhotra, Andres González-Bonet, Seok-Hee Hyun, Elaine Moase, Theresa M Allen, David H Thompson
ABSTRACT
A family of 3-methoxypoly(ethylene glycol)-vinyl ether-1,2-dioleylglycerol (mPEG-VE-DOG) lipopolymer conjugates, designed on the basis of DFT calculations to possess a wide range of proton affinities, was synthesized and tested for their hydrolysis kinetics in neutral and acidic buffers. Extruded ∼100 nm liposomes containing these constructs in ≥90 mol % 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE) produced dispersions that retained their calcein cargo for more than 2 days at pH 7.5, but released the encapsulated contents over a wide range of time scales as a function of the electronic properties of the vinyl ether linkage, the solution pH, and the mPEG-VE-DOG composition in the membrane. The in vivo performance of two different 90:10 DOPE:mPEG-VE-DOG compositions was also evaluated for blood circulation time and biodistribution in mice, using (125)I-tyraminylinulin as a label. The pharmacokinetic profiles gave a t(1/2) of 7 and 3 h for 90:10 DOPE:ST302 and 90:10 DOPE:ST502, respectively, with the liposomes being cleared predominantly by liver and spleen uptake. The behavior of these DOPE:mPEG-VE-DOG formulations is consistent with their relative rates of vinyl ether hydrolysis, i.e., the more acid-sensitive mPEG-VE-DOG derivatives produced faster leakage rates from DOPE:mPEG-VE-DOG liposomes, but decreased the blood circulation times in mice. These findings suggest that the vinyl ether-based PEG-lipid derivatives are promising agents for stabilizing acid-sensitive DOPE liposomes to produce formulations with a priori control over their pH responsiveness in vitro. Our data also suggest, however, that the same factors that contribute to enhanced acid sensitivity of the DOPE:mPEG-VE-DOG dispersions are also likely responsible for their reduced pharmacokinetic profiles.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Pricing and availability is not currently available.
Sigma-Aldrich
Dioleoylglycerol, ≥99%, mixture of 1,3- and 1,2-isomers, liquid
Pricing and availability is not currently available.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Pricing and availability is not currently available.