Merck
  • A functionalized poly(amidoamine) nanocarrier-loading 5-fluorouracil: pH-responsive drug release and enhanced anticancer effect.
A functionalized poly(amidoamine) nanocarrier-loading 5-fluorouracil: pH-responsive drug release and enhanced anticancer effect.
Anti-cancer drugs (2012-09-20)
Lina Du, Yiguang Jin, Jiangyong Yang, Shuangmiao Wang, Xiangtao Wang
ABSTRACT
A functionalized poly(amidoamine) (PAMAM) nanocarrier was designed and prepared to deliver anticancer drugs. The nanocarrier is a copolymer with a core-shell structure with 3.0 G PAMAM as the core and sequentially conjugated poly(2-(N,N-diethylamino)ethyl methacrylate) (pDEA) and methoxy-poly(ethylene glycol) 2000 (mPEG) as the shell. The copolymer, PAMAM-pDEA-mPEG, was synthesized using atom transfer radical polymerization and click chemistry. The PAMAM core loaded drugs. pDEA had pH-sensitive properties, showing hydrophobicity in neutral environments and hydrophilicity in weakly acidic environments because of the presence of tertiary amines. Therefore, pDEA was a functional layer coating drugs in neutral environments and releasing drugs in acidic environments. The outer mPEG layer allowed the nanocarrier to circulate in the blood for a long period of time and improved the stability of the nanocarriers. The anticancer drug 5-fluorouracil (5-FU) was entrapped in the nanocarrier at high levels by changing the pH from 4.0 to 8.0. The drug release was also highly pH responsive, and the release rate was much higher at pH 6.5 than at pH 7.4, which favored drug release in the weakly acidic tumor environment. The blank nanocarrier was not toxic to cells or animals. The 5-FU-loaded nanocarrier exerted enhanced anticancer effects on tumor-bearing mice relative to 5-FU alone. PAMAM-pDEA-mPEG is a promising nanocarrier for the delivery of anticancer drugs.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 20,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 10,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 550
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn ~2,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 750
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, BioUltra, 500
Pricing and availability is not currently available.
Sigma-Aldrich
Poly(ethylene glycol) methyl ether, average Mn 5,000
Pricing and availability is not currently available.
Sigma-Aldrich
Methoxypolyethylene glycol 350, average mol wt 350
Pricing and availability is not currently available.