Merck
  • A supramolecular complex in small-molecule solar cells based on contorted aromatic molecules.
A supramolecular complex in small-molecule solar cells based on contorted aromatic molecules.
Angewandte Chemie (International ed. in English) (2012-07-19)
Seok Ju Kang, Jong Bok Kim, Chien-Yang Chiu, Seokhoon Ahn, Theanne Schiros, Stephanie S Lee, Kevin G Yager, Michael F Toney, Yueh-Lin Loo, Colin Nuckolls
ABSTRACT
"Ball and socket" motif: The contorted dibenzotetrathienocoronene (6-DBTTC) forms a complex with the C(70) fullerene PC(70) BM embedded in an amorphous phase of PC(70) BM. The materials are processable into organic solar cells in solution. The power conversion efficiency is maximal when there is a 1:2 molar ratio of 6-DBTTC to PC(70) BM. Formation of the supramolecular complex directly affects charge separation in the active layer.
Sigma-Aldrich
Coronene, purified by sublimation, 99%
Coronene, BCR®, certified reference material
Sigma-Aldrich
Coronene, 97%