Merck
  • Genome sequence of Escherichia coli J53, a reference strain for genetic studies.
Genome sequence of Escherichia coli J53, a reference strain for genetic studies.
Journal of bacteriology (2012-06-29)
Hana Yi, Yong-Joon Cho, Dongeun Yong, Jongsik Chun
ABSTRACT
Escherichia coli J53 (F(-) met pro Azi(r)) is a derivative of E. coli K-12 which is resistant to sodium azide. This strain has been widely used as a general recipient strain for various conjugation experiments. Here, we report the genome sequence of E. coli J53 (=KACC 16628).
Sigma-Aldrich
Sodium azide 0.1 M solution
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)