Merck
  • Scalable synthesis of enantiomerically pure bicyclo[2.2.2]octadiene ligands.
Scalable synthesis of enantiomerically pure bicyclo[2.2.2]octadiene ligands.
The Journal of organic chemistry (2012-05-04)
Stefan Abele, Roman Inauen, Dirk Spielvogel, Christian Moessner
ABSTRACT
An operationally simple and scalable synthesis of enantiomerically pure bicyclo[2.2.2]octadiene (bod*) ligands relying on an organocatalytic one-pot Michael addition-aldol reaction with cheap 2-cyclohexenone and phenylacetaldehyde is presented. The crystalline bicyclic product 4a (6-hydroxy-5-phenylbicyclo[2.2.2]octan-2-one) is transformed into phenylbicyclo[2.2.2]oct-5-en-2-one 2, a versatile starting material for the 2-step synthesis of both symmetrical, such as Hayashi's Ph-bod* ligand, as well as novel unsymmetrical chiral dienes.
Sigma-Aldrich
1-Naphthylmagnesium bromide solution, 0.25 M slurry in THF
Sigma-Aldrich
Phenylacetaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Phenylacetaldehyde, ≥90%
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%