Merck
A convergent approach to polycyclic aromatic hydrocarbons.
Chemical communications (Cambridge, England) (2011-10-15)
Raphaël F Guignard, Samir Z Zard
ABSTRACT
A new concise route to Polycyclic Aromatic Hydrocarbons (PAHs) through radical addition and cyclisation of xanthates is described.
Sigma-Aldrich
Acetophenone, ≥98%, FG
Supelco
Acetophenone, analytical standard
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Acetophenone, puriss. p.a., ≥99.0% (GC)