Merck
  • A practical, one-pot synthesis of highly substituted thiophenes and benzo[b]thiophenes from bromoenynes and o-alkynylbromobenzenes.
A practical, one-pot synthesis of highly substituted thiophenes and benzo[b]thiophenes from bromoenynes and o-alkynylbromobenzenes.
Organic letters (2011-09-03)
Verónica Guilarte, Manuel A Fernández-Rodríguez, Patricia García-García, Elsa Hernando, Roberto Sanz
ABSTRACT
An efficient synthesis of thiophenes and benzo[b]thiophenes has been developed from easily available bromoenynes and o-alkynylbromobenzene derivatives. This novel one-pot procedure involves a Pd-catalyzed C-S bond formation using a hydrogen sulfide surrogate followed by a heterocyclization reaction. Moreover, in situ functionalization with selected electrophiles further expands the potential of this methodology to the preparation of the corresponding highly substituted sulfur heterocycles.
Sigma-Aldrich
Bromobenzene, ReagentPlus®, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Bromobenzene, ≥99.5% (GC)
Pricing and availability is not currently available.