Merck
  • Allergy to methyldibromoglutaronitrile/phenoxyethanol (Euxyl k 400): regulatory issues, epidemiology, clinical characteristics, and management.
Allergy to methyldibromoglutaronitrile/phenoxyethanol (Euxyl k 400): regulatory issues, epidemiology, clinical characteristics, and management.
Dermatitis : contact, atopic, occupational, drug (2011-05-17)
Angela E Aakhus, Erin M Warshaw
ABSTRACT
Methyldibromoglutaronitrile/phenoxyethanol (Euxyl K 400) is a preservative found in both personal care products and industrial sources. Although Euxyl K 400 initially appeared to have low sensitizing potential, increased prevalence of contact allergy to Euxyl K 400 led to regulatory intervention. This review summarizes the history, epidemiology, and management of contact allergy to Euxyl K 400. Issues related to patch-test preparations are also discussed.
Product Description
Supelco
2-Phenoxyethanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methyldibromoglutaronitrile, analytical standard
Supelco
2-Phenoxyethanol, analytical standard
Sigma-Aldrich
2-Phenoxyethanol, ≥99%
USP
Phenoxyethanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylene glycol monophenyl ether, ≥90%
Phenoxyethanol, European Pharmacopoeia (EP) Reference Standard