Merck
  • Characterizing workplace exposures in Vietnamese women working in California nail salons.
Characterizing workplace exposures in Vietnamese women working in California nail salons.
American journal of public health (2011-05-10)
Thu Quach, Robert Gunier, Alisha Tran, Julie Von Behren, Phuong-An Doan-Billings, Kim-Dung Nguyen, Linda Okahara, Benjamin Yee-Bun Lui, Mychi Nguyen, Jessica Huynh, Peggy Reynolds
ABSTRACT
We engaged Vietnamese nail salon workers in a community-based participatory research (CBPR) study to measure personal and area concentrations of solvents in their workplace. We measured average work-shift concentrations of toluene, ethyl acetate, and isopropyl acetate among 80 workers from 20 salons using personal air monitors. We also collected area samples from 3 salons using summa canisters. For personal measurements, the arithmetic mean was 0.53 parts per million (range = 0.02-5.50) for ethyl acetate, 0.04 parts per million (range = 0.02-0.15) for isopropyl acetate, and 0.15 parts per million (range = 0.02-1.0) for toluene. Area measurements were lower in comparison, but we detected notable levels of methyl methacrylate, a compound long banned from nail products. Predictors of solvent levels included different forms of ventilation and whether the salon was located in an enclosed building. Using a CBPR approach that engaged community members in the research process contributed to the successful recruitment of salon workers. Measured levels of toluene, methyl methacrylate, and total volatile organic compounds were higher than recommended guidelines to prevent health symptoms such as headaches, irritations, and breathing problems, which were frequently reported in this workforce.
Sigma-Aldrich
Isopropyl acetate, ≥99%, FCC
Supelco
Isopropyl acetate, analytical standard
Sigma-Aldrich
Isopropyl acetate, 98%
Sigma-Aldrich
Isopropyl acetate, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Isopropyl acetate, ≥99.6%