Merck
  • Microwave-assisted regioselective ring opening of non-activated aziridines by lithium aluminium hydride.
Microwave-assisted regioselective ring opening of non-activated aziridines by lithium aluminium hydride.
Organic & biomolecular chemistry (2010-08-05)
Sonja Stanković, Matthias D'hooghe, Norbert De Kimpe
ABSTRACT
A new synthetic protocol for the LiAlH(4)-promoted reduction of non-activated aziridines under microwave conditions was developed. Thus, ring opening of 2-(acetoxymethyl)aziridines provided the corresponding beta-amino alcohols, which were then used as eligible substrates in the synthesis of 5-methylmorpholin-2-ones via condensation with glyoxal in THF. The same procedure was applied for the preparation of novel 5(R)- and 5(S)-methylmorpholin-2-ones starting from the corresponding enantiopure 2-(hydroxymethyl)aziridines. Additionally, 2-(methoxymethyl)- and 2-(phenoxymethyl)aziridines were treated with LiAlH(4) under microwave irradiation, giving rise to either isopropylamines or 1-methoxypropan-2-amines depending on the reaction conditions.
Sigma-Aldrich
Lithium aluminum hydride, hydrogen-storage grade
Sigma-Aldrich
Lithium aluminum hydride, pellets, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Lithium aluminum hydride solution, 0.5 M in 2-methoxyethyl ether
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric), tablet (5 g each)
Sigma-Aldrich
Lithium aluminum hydride solution, 1.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride solution, 2.0 M in THF
Sigma-Aldrich
Lithium aluminum hydride, ≥97.0% (gas-volumetric)
Sigma-Aldrich
Lithium aluminum hydride, powder, reagent grade, 95%
Sigma-Aldrich
Lithium aluminum hydride solution, 2.3 M in 2-methyltetrahydrofuran