Merck
  • Biosynthesis of lovastatin and related metabolites formed by fungal iterative PKS enzymes.
Biosynthesis of lovastatin and related metabolites formed by fungal iterative PKS enzymes.
Biopolymers (2010-06-26)
Chantel D Campbell, John C Vederas
ABSTRACT
The fungal polyketide lovastatin is a cholesterol lowering agent that is an immediate precursor to a multi-billion dollar drug, simvastatin (Zocor). Lovastatin is produced by an iterative type I polyketide synthase known as LovB and a partner enoyl reductase (LovC). There is evidence that a Diels-Alderase enzyme activity is utilized in its biosynthesis. This review examines the biosynthesis of lovastatin, as well as of compactin, equisetin, cytochalasins, and solanapyrones, which are other structurally related polyketides that appear to utilize a Diels-Alderase.
Product Description
Lovastatin for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mevinolin from Aspergillus sp., ≥98% (HPLC)
Sigma-Aldrich
Mevastatin, ≥98% (HPLC), powder or crystals
USP
Lovastatin, United States Pharmacopeia (USP) Reference Standard
Supelco
Lovastatin, Pharmaceutical Secondary Standard; Certified Reference Material
Lovastatin, European Pharmacopoeia (EP) Reference Standard