Merck
  • Chemoprevention of colorectal cancer by targeting APC-deficient cells for apoptosis.
Chemoprevention of colorectal cancer by targeting APC-deficient cells for apoptosis.
Nature (2010-03-30)
Ling Zhang, Xiaoyang Ren, Eckhard Alt, Xiaowen Bai, Shaoyi Huang, Zhengming Xu, Patrick M Lynch, Mary P Moyer, Xian-Feng Wen, Xiangwei Wu
ABSTRACT
Cancer chemoprevention uses natural, synthetic, or biological substances to reverse, suppress, or prevent either the initial phase of carcinogenesis or the progression of neoplastic cells to cancer. It holds promise for overcoming problems associated with the treatment of late-stage cancers. However, the broad application of chemoprevention is compromised at present by limited effectiveness and potential toxicity. To overcome these challenges, here we developed a new chemoprevention approach that specifically targets premalignant tumour cells for apoptosis. We show that a deficiency in the adenomatous polyposis coli (APC) gene and subsequent activation of beta-catenin lead to the repression of cellular caspase-8 inhibitor c-FLIP (also known as CFLAR) expression through activation of c-Myc, and that all-trans-retinyl acetate (RAc) independently upregulates tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) death receptors and suppresses decoy receptors. Thus, the combination of TRAIL and RAc induces apoptosis in APC-deficient premalignant cells without affecting normal cells in vitro. In addition, we show that short-term and non-continuous TRAIL and RAc treatment induce apoptosis specifically in intestinal polyps, strongly inhibit tumour growth, and prolong survival in multiple intestinal neoplasms C57BL/6J-Apc(Min)/J (Apc(Min)) mice. With our approach, we further demonstrate that TRAIL and RAc induce significant cell death in human colon polyps, providing a potentially selective approach for colorectal cancer chemoprevention by targeting APC-deficient cells for apoptosis.
Product Description
Supelco
Retinyl Acetate (Vitamin A Acetate), Pharmaceutical Secondary Standard; Certified Reference Material
Retinol acetate, European Pharmacopoeia (EP) Reference Standard
Supelco
Retinyl acetate, analytical standard
Sigma-Aldrich
Retinyl acetate, solid or viscous liquid, BioReagent, synthetic, suitable for cell culture
Sigma-Aldrich
Retinyl acetate, synthetic, crystalline solid or supercooled liquid
Sigma-Aldrich
Retinyl acetate, synthetic, matrix dispersion, 475,000-650,000 USP units/g